Phenylphosphinic acid 99%

Danger

AC13078-0050
200050-456EA 22.1 USD
200050-456
Phenylphosphinic acid 99%
Phenylphosphinic acid
Formula: C₆H₇O₂P
Boiling Pt: 180 °C
Melting Pt: 82…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002131
CAS Number: 1779-48-2
EINECS: 217-217-3
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White Crystalline solid
Infrared spectrum Conforms
Melting point 82 to 87 °C
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR