DL-α-Phenylglycine 99%

Synonyms: H-DL-Phg-OHDL-2-PhenylglycineDl-A-Aminophenylacetic Acid

AC13064-1000
200025-178EA 37.4 USD
200025-178
DL-α-Phenylglycine 99%
DL-α-Phenylglycine
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Melting Pt: 290 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00064402
CAS Number: 2835-06-5
EINECS: 220-608-1

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Appearance of solution (5% in 1N HCl) clear colorless to slightly yellow
Titration with HClO4 ≥98.5 %
UV at 200 nm A: ≤0.010
Gradient analysis Passes test
Absorbance Passes test
KMnO4-reducing substance passes test
Conductivity 2.0 x 10-6 ohm-1 cm-1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR