o-Phenylenediacetonitrile 99+%

Warning

Synonyms: 1,2-Phenylenediacetonitrile1,2-Bis(cyanomethyl)-benzene

AC13052-0100 AC13052-0500
200045-692EA 40.2 USD
200045-692 200045-694
o-Phenylenediacetonitrile 99+%
o-Phenylenediacetonitrile
Formula: C₁₀H₈N₂
MW: 156.19 g/mol
Melting Pt: 58…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00001905
CAS Number: 613-73-0
EINECS: 210-351-3
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow to beige Crystalline powder or crystals (May contain yellow to orange chunks)
Infrared spectrum Conforms
Melting point 57 to 61 °C
GC ≥99.0 %
Residue after evaporation ≤0.001 %
Filtered to 0.2 micrometer Filtered

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR