Perfluorodecahydronaphthalene (mixture of cis and trans isomers) cis- and trans-mixture 90%

Warning

Synonyms: Perfluoro(decahydronaphthalene)PerfluorodecahydronaphthalenePerfluorodecalin

AC13004-1000 AC13004-0250
200013-940EA 53.2 USD
200013-940 103525-352
Perfluorodecahydronaphthalene (mixture of cis and trans isomers) cis- and trans-mixture 90%
Perfluorodecahydronaphthalene (mixture of cis and trans isomers)
Formula: C₁₀F₁₈
MW: 462.08 g/mol
Boiling Pt: 142 °C (760 mmHg)
Melting Pt: –10 °C
Density: 1.9 g/cm³
Flash Pt: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD00010626
CAS Number: 306-94-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR