Trichloromethanesulfenyl chloride 97%

Danger

Synonyms: Perchloromethyl mercaptan

AC13003-0250 AC13003-0050
200020-332EA 93.7 USD
200020-332 103530-886
Trichloromethanesulfenyl chloride 97%
Trichloromethanesulphenyl chloride
Formula: CCl₄S
Boiling Pt: 146…148 °C (760 mmHg)
Density: 1.7 g/cm³
Storage Temperature: Ambient
EINECS: 209-840-4
UN: 1670
ADR: 6.1,I

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5370 to 1.5410 (20 °C, 589 nm)
Assay ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR