D-(-)-Penicillamine 99%, meets analytical specification of Ph. Eur., USP
|
Danger
|
Synonyms:
H-D-Pen-OH
, (S)-2-Amino-3-mercapto-3-methylbutanoic acid
AC12974-0250
AC12974-0050
200030-846EA
101
USD
200030-846
200043-072
D-(-)-Penicillamine 99%, meets analytical specification of Ph. Eur., USP
D(-)-Penicillamine
Formula:
C₅H₁₁NO₂S MW: 149.2 g/mol Melting Pt: 212 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00064302 CAS Number: 52-67-5 EINECS: 200-148-8 |
Specification Test Results
Appearance | White to almost white Powder |
Infrared spectrum | Conforms |
Assay | 97.0 to 102.0 % (USP) |
Loss on drying | ≤0.5 % (60 °C, 3 h) (vacuum) |
UV | at 268 nm A: ≤0.07 (0.1 g in 50 ml water) |
Appearance of solution | Passes test |
Identification A | Passes test |
Identification B | Passes test |
Identification C | Passes test |
Identification D | Passes test |
Residue after ignition | ≤0.1 % |
pH | 4.5 to 5.5 (1 g in 100 ml water) |
Specific optical rotation | -60.5° to -65.0° (20 °C, 589 nm) (0.5g in 10 ml 1N NaOH) |
Titration with HClO4 | 98.0 to 101.0 % (EP) |
Mercury (Hg) | ≤0.001 % |
Impurity | ≤1 % (Penicillamine disulfide) |
Impurity | ≤0.1 % (Penicillin) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...