2-Oxazolidinone 98%

Synonyms: Oxazolidin-2-one2-Oxazolidone

AC12966-1000 AC12966-0250
200022-898EA 126 USD
200022-898 200009-560
2-Oxazolidinone 98%
2-Oxazolidinone
Formula: C₃H₅NO₂
MW: 87.08 g/mol
Boiling Pt: 220 °C (48 mmHg)
Melting Pt: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00005268
CAS Number: 497-25-6
EINECS: 207-840-9

Order Now

Specification Test Results

Appearance Off-white to yellow to light beige Powder or crystals
Infrared spectrum Conforms
GC ≥97.5 %
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR