Norbornene ≥99% stabilized

Supplier: ACROS ORGANICS MS

Danger

Synonyms: Norbornylene

AC12924-0025 AC12924-5000 AC12924-0250
103524-088EA 232.48 USD
103524-088 200013-722 200007-198
Norbornene ≥99% stabilized
Norbornene
Formula: C₇H₁₀
MW: 94.16 g/mol
Boiling Pt: 96 °C (760 mmHg)
Melting Pt: 42…46 °C
Flash Pt: –15 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00082304
CAS Number: 498-66-8
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Appearance White Crystalline mass
Infrared spectrum Conforms
Melting point 42°C to 46°C
GC ≥98.5 %
Stabilizer 0.1 % phenolic antioxidant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR