Nipecotamide 95%
|
Warning
|
Synonyms:
Piperidine-3-carboxamide
AC12830-0050
AC12830-2500
AC12830-0500
103510-162EA
2058.5
USD
103510-162
200020-084
200032-256
Nipecotamide 95%
Nipecotamide
Formula:
C₆H₁₂N₂O MW: 128.1700 g/mol Melting Pt: 102…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005993 CAS Number: 4138-26-5 EINECS: 223-962-5 |
Specification Test Results
Appearance | Yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 102 to 108 °C |
Titration with HClO4 | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...