Myrcene 90% tech. stabilized

Danger

AC12808-5000 AC12808-0025 AC12808-0050
200013-430EA 74.4 USD
200013-430 103524-066 200032-012
Myrcene 90% tech. stabilized
Myrcene
Formula: C₁₀H₁₆
Boiling Pt: 165 °C (760 mmHg)
Density: 0.795 g/cm³
Flash Pt: 44 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008908
CAS Number: 123-35-3
EINECS: 204-622-5
UN: 2319
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4660 to 1.4720 (20 °C, 589 nm)
Density (20/4°C) 0.789 to 0.800 g/ml
GC ≥88 %
Stabilizer ≤0.15 % alpha-Tocopherol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR