trans,trans-Muconic acid (trans,trans-1,3-butadiene-1,4-dicarboxylic acid) 97%
|
Warning
|
Synonyms:
trans, trans-2,4-Hexadienedioic acid
AC12807-0050
AC12807-0010
103510-152EA
412
USD
103510-152
200007-910
trans,trans-Muconic acid (trans,trans-1,3-butadiene-1,4-dicarboxylic acid) 97%
trans,trans-Muconic acid (trans,trans-1,3-Butadiene-1,4-dicarboxylic acid)
Formula:
C₆H₆O₄ MW: 142.11 g/mol Boiling Pt: 320 °C (760 mmHg) Melting Pt: 290 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002702 CAS Number: 3588-17-8 EINECS: 222-724-8 |
Specification Test Results
Appearance | Beige Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Titration with NaOH | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...