3-Methyl-1-phenyl-2-pyrazolin-5-one 99%
|
Warning
|
Synonyms:
1-Phenyl-3-methyl-5-pyrazolone
, 3-Methyl-1-phenyl-1H-pyrazol-5(4H)-one
, MCI-186
, Edaravone
AC12746-0050
AC12746-5000
AC12746-1000
200010-016EA
56.9
USD
200010-016
200041-636
200017-142
3-Methyl-1-phenyl-2-pyrazolin-5-one 99%
3-Methyl-1-phenyl-2-pyrazolin-5-one
Formula:
C₁₀H₁₀N₂O MW: 174.2 g/mol Boiling Pt: 287 °C (265 mmHg) Melting Pt: 127…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003138 CAS Number: 89-25-8 EINECS: 201-891-0 |
Specification Test Results
Appearance | Yellow to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 127 to 131 °C |
HPLC | ≥98.5 % |
Water adsorption | ≥20 % (50% rH, 1 bar, 25 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...