Methylmalonic acid 96%

Warning

Synonyms: 2-Methylmalonic acid

AC12709-1000 AC12709-0250 AC12709-0050
103510-140EA 517 USD
103510-140 200049-938 200025-914
Methylmalonic acid 96%
Methylmalonic acid
Formula: C₄H₆O₄
MW: 118.09 g/mol
Melting Pt: 129…136 °C
Density: 1.45 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00002656
CAS Number: 516-05-2
EINECS: 208-219-5

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 129 to 136 °C
Water ≤0.5 % (K.F.)
Titration with NaOH ≥95 %
Color scale ≤100 APHA
Acidity ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR