N-Methyliminodiacetic acid 99%

Warning

Synonyms: 2,2'-(Methylazanediyl)diacetic acid

AC12701-0050 AC12701-0250
200026-210EA 52.71 USD
200026-210 200032-292
N-Methyliminodiacetic acid 99%
N-Methyliminodiacetic acid
Formula: C₅H₉NO₄
MW: 147.13 g/mol
Melting Pt: 218…221 °C
Storage Temperature: Ambient
EINECS: 224-557-6

Order Now

Specification Test Results

Appearance White to cream Powder
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR