Methyl-2-hydroxyisobutyrate 99%
|
Warning
|
Synonyms:
2-Hydroxyisobutyric Acid Methyl Ester
, Methyl 2-Hydroxyisobutyrate
AC12697-0050
AC12697-0500
200049-860EA
34
USD
200049-860
200031-840
Methyl-2-hydroxyisobutyrate 99%
Methyl-2-hydroxyisobutyrate
Appearance: Clear colorless Liquid
Formula:
C₅H₁₀O₃ MW: 118.13 g/mol Boiling Pt: 137 °C Density: 1.03 g/cm³ Flash Pt: 41 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004457 CAS Number: 2110-78-3 UN: 3272 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4100 to 1.4120 (20 °C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...