N-Methylphenethylamine 99%

Danger

AC12745-0050 AC12745-0250 AC12745-1000
200020-130EA 83.8 USD
200020-130 103530-824 200038-260
N-Methylphenethylamine 99%
N-Methylphenethylamine
Formula: C₉H₁₃N
Boiling Pt: 203 °C (760 mmHg)
Density: 0.93 g/cm³
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00008291
EINECS: 209-632-3
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5150 to 1.5170 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR