N-Methylformanilide 99%

Warning

AC12681-0100 AC12681-5000 AC12681-0010 AC12681-1000
103466-166EA 875 USD
103466-166 200032-290 200038-246 200013-704
N-Methylformanilide 99%
N-Methylformanilide
Formula: C₈H₉NO
Boiling Pt: 243…244 °C (760 mmHg)
Melting Pt: 8…13 °C
Density: 1.09 g/cm³
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00003283
CAS Number: 93-61-8
EINECS: 202-262-3

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5600 to 1.5620 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR