4,4'-Bis(dimethylamino)diphenylmethane 98%

Danger

Synonyms: 4,4'-Methylenebis(N,N-dimethylaniline)Bis[4-(dimethylamino)phenyl]methane

AC12674-1000 AC12674-0050 AC12674-5000
200023-500EA 77.5 USD
200023-500 200043-978 200029-494
4,4'-Bis(dimethylamino)diphenylmethane 98%
4,4'-Bis(dimethylamino)diphenylmethane
Formula: C₁₇H₂₂N₂
MW: 254.38 g/mol
Melting Pt: 86…90 °C
Flash Pt: 178 °C
Storage Temperature: Ambient
MDL Number: MFCD00008317
CAS Number: 101-61-1
EINECS: 202-959-2
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to beige to grey to brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 86 to 92 °C
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR