Methyl cyclopropanecarboxylate 98%

Danger

Synonyms: Methylcyclopropanecarboxylate

AC12671-0250 AC12671-1000 AC12671-5000
103530-802EA 488 USD
103530-802 200013-286 200049-904
Methyl cyclopropanecarboxylate 98%
Methyl cyclopropanecarboxylate
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 119 °C (764 mmHg)
Density: 0.98 g/cm³
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00001280
CAS Number: 2868-37-3
EINECS: 220-690-9
UN: 1992
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4170 to 1.4190 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR