Methyl-4-bromocrotonate 85% tech.

Danger

Synonyms: Methyl 4-bromobut-2-enoate

AC12645-0250 AC12645-1000 AC12645-5000
200013-242EA 88.2 USD
200013-242 200031-866 103530-792
Methyl-4-bromocrotonate 85% tech.
Methyl 4-bromocrotonate
Formula: C₅H₇BrO₂
MW: 179.01 g/mol
Boiling Pt: 83…85 °C (13 mmHg)
Density: 1.52 g/cm³
Flash Pt: 91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000246
EINECS: 214-251-0
UN: 1760
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4970 to 1.5050 (20 °C, 589 nm)
GC ≥80 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR