5-Methoxytryptamine hydrochloride 97%
|
Warning
|
Synonyms:
2-(5-Methoxy-1H-indol-3-yl)ethanamine
, O-Methylserotonin hydrochloride
AC12613-2500
AC12613-0010
200042-320EA
67.6
USD
200042-320
200024-108
5-Methoxytryptamine hydrochloride 97%
5-Methoxytryptamine hydrochloride
Formula:
C₁₁H₁₅ClN₂O Melting Pt: 246…248 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005662 CAS Number: 66-83-1 EINECS: 200-637-6 |
Specification Test Results
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤0.5 % |
Loss on drying | ≤1 % |
Titration with NaOH | ≥96.0 % |
Titration with HClO4 | ≥98.0 % |
Insoluble matter | ≤0.005 % (in water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...