1-(3-Methoxyphenyl)piperazine dihydrochloride 98%
|
Warning
|
AC12605-0250
200027-476EA
207
USD
200027-476
1-(3-Methoxyphenyl)piperazine dihydrochloride 98%
1-(3-Methoxyphenyl)piperazine dihydrochloride
Formula:
C₁₁H₁₈Cl₂N₂O Melting Pt: 201…204 °C Storage Temperature: Ambient |
MDL Number:
MFCD00035285 CAS Number: 6968-76-9 |
Specification Test Results
Appearance | White to grey to pink Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Titration with HClO4 | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...