5-Methoxyindole-2-carboxylic acid 99%

AC12590-0100
200017-850EA 120.3 USD
200017-850
5-Methoxyindole-2-carboxylic acid 99%
5-Methoxyindole-2-carboxylic acid

Appearance: Light yellow to brown Powder

Formula: C₁₀H₉NO₃
Melting Pt: 197…201 °C
Storage Temperature: Ambient
MDL Number: MFCD00005614
CAS Number: 4382-54-1

Order Now

Specification Test Results

Appearance Light yellow to brown Powder
Infrared spectrum Conforms
Melting point 197 to 201 °C
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR