Menthone (mixture of isomers) 98%

Warning

Synonyms: 2-Isopropyl-5-Methylcyclohexanone

AC12541-1000
200013-142EA 111 USD
200013-142
Menthone (mixture of isomers) 98%
Menthone (mixture of isomers)
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Melting Pt: –6 °C
Density: 0.89 g/cm³
Flash Pt: 69 °C
Storage Temperature: Ambient
MDL Number: MFCD00062998
CAS Number: 10458-14-7
EINECS: 233-944-9

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4460 to 1.4550 (20 °C, 589 nm)
Density (20 °C) 0.889 to 0.898 g/ml
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR