N-(Diphenylmethylene)glycine ethyl ester 98%

Synonyms: Ethyl N-(diphenylmethylene)glycinateEthyl 2-((diphenylmethylene)amino)acetateEthyl (diphenylmethylenamino)acetateEthyl-2-((diphenylmethylene)amino)acetate

AC12413-0050 AC12413-0250
200012-420EA 64.99 USD
200012-420 103510-094
N-(Diphenylmethylene)glycine ethyl ester 98%
N-(Diphenylmethylene)glycine ethyl ester
Formula: C₁₇H₁₇NO₂
MW: 267.32 g/mol
Melting Pt: 51…55 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010590
CAS Number: 69555-14-2

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 48 to 54 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR