Oxytetracycline dihydrate 95%

Warning

AC12384-1000 AC12384-0100
200020-284EA 355 USD
200020-284 103466-258
Oxytetracycline dihydrate 95%
Oxytetracycline dihydrate
Formula: C₂₂H₂₄N₂O₉·2H₂O
Melting Pt: 181…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00151234
CAS Number: 6153-64-6

Order Now

Specification Test Results

Appearance Yellow to beige Crystalline powder
Infrared spectrum Conforms
Water 6 to 9 % (K.F.)
HPLC 94 to 102 % (on anhydrous substance)
Specific optical rotation -203° to -216° (20 °C, 589 nm) (c=1, 0.1 M HCl)
Specific optical rotation on anhydrous substance

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR