1-(2-Bromoethoxy)-4-fluorobenzene 95%
|
Warning
|
Synonyms:
4-Fluorophenoxy-ethylbromide
AC12416-0250
200013-952EA
143.1
USD
200013-952
1-(2-Bromoethoxy)-4-fluorobenzene 95%
1-(2-Bromoethoxy)-4-fluorobenzene
Formula:
C₈H₈BrFO Storage Temperature: Ambient |
MDL Number:
MFCD00044739 CAS Number: 332-48-9 |
Specification Test Results
Appearance | Clear colorless to brown Liquid after melting |
Infrared spectrum | Conforms |
Refractive index | 1.5300 to 1.5320 (20 °C, 589 nm) |
GC | ≥94 % |
Color scale | ≤30 APHA |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...