Diethyl (2-oxopropyl)phosphonate 96%
|
Warning
|
Synonyms:
Diethyl acetylmethylphosphonate
AC12369-0050
200012-062EA
88.4
USD
200012-062
Diethyl (2-oxopropyl)phosphonate 96%
Diethyl (2-oxopropyl)phosphonate
Formula:
C₇H₁₅O₄P MW: 194.17 g/mol Boiling Pt: 98 °C (1 mmHg) Density: 1.11 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044728 CAS Number: 1067-71-6 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4325 to 1.4355 (20 °C, 589 nm) |
GC | ≥95 % |
Iron (Fe) | ≤0.0001 % |
Heavy metals (as Pb) | ≤1 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...