Hesperidin 97% includes analogeous compounds

Synonyms: (S)-5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)chroman-4-one

AC12346-0050 AC12346-2500
200019-302EA 17 USD
200019-302 200031-514
Hesperidin 97% includes analogeous compounds
Hesperidin
Formula: C₂₈H₃₄O₁₅
MW: 610.56 g/mol
Melting Pt: 258…262 °C
Storage Temperature: Ambient
EINECS: 208-288-1

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
Loss on drying ≤4 %
Heavy metals ≤20 ppm
Additional info residue by calcination: < 0.3%
UV spectrophotometry ≥96.0 % (on dry substance)
Volatile matter ≤5.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR