Ethyl-2-oxocyclopentaneacetate 95%

Synonyms: Ethyl 2-(2-Oxocyclopentyl)acetate2-(2-Oxocyclopentyl)acetic Acid Ethyl EsterEthyl 2-oxocyclopentylacetateEthyl 2-(2-oxocyclopentyl)acetate2-Oxocyclopentaneacetic Acid Ethyl Ester

AC12304-0010 AC12304-0050
200012-460EA 116 USD
200012-460 103510-078
Ethyl-2-oxocyclopentaneacetate 95%
Ethyl-2-oxocyclopentaneacetate
Formula: C₉H₁₄O₃
MW: 170.21 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00044715
CAS Number: 20826-94-2

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94 %
GC ≥94 %
Specific gravity 0.859 to 0.864
Specific optical rotation -15° to -21° (20 °C, 589 nm) (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR