4-Phenylpiperidine-4-carboxylic acid 4-toluenesulfonate 98%

Warning

AC12292-1000 AC12292-0250
200010-612EA 144 USD
200010-612 200017-690
4-Phenylpiperidine-4-carboxylic acid 4-toluenesulfonate 98%
4-Phenyl-4-piperidinecarboxylic acid p-methylbenzenesulfonate

Appearance: Off-white to light beige Crystalline powder

Formula: C₁₂H₁₅NO₂·C₇H₈O₃S
MW: 377.46 g/mol
Melting Pt: 225…229 °C
Storage Temperature: Ambient
MDL Number: MFCD00044709
CAS Number: 83949-32-0

Order Now

Specification Test Results

Appearance Off-white to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 225 to 229 °C
Water ≤1 % (K.F.)
Solubility (3 % in boiling water) Clear colorless to almost colorless
Titration with NaOH 97.5 to 102.5 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR