Isonipecotamide 98%

Warning

Synonyms: Piperidine-4-carboxamide

AC12261-1000
200006-730EA 73.7 USD
200006-730
Isonipecotamide 98%
Isonipecotamide
Formula: C₆H₁₂N₂O
MW: 128.18 g/mol
Melting Pt: 146…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00038012
CAS Number: 39546-32-2
EINECS: 254-501-6

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 146 to 150 °C (on dry substance)
Water ≤2 % (K.F.)
Loss on drying ≤2 % (70 °C)
Sulfated ash ≤0.2 %
Titration with HClO4 ≥97.5 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR