Isobutylamine

Danger

Synonyms: 1-Amino-2-methylpropane2-Methylpropylamine

AC12250-0010 AC12250-0025 AC12250-2500
102774-854EA 74.6 USD
102774-854 102775-938 102773-914
Isobutylamine
Isobutylamine
Formula: C₄H₁₁N
MW: 73.14 g/mol
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: –85 °C
Density: 0.73 g/cm³
Flash Pt: –9 °C
Storage Temperature: Ambient
MDL Number: MFCD00008146
CAS Number: 78-81-9
UN: 1214
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3960 to 1.3980 (20 °C, 589 nm)
GC ≥98.5 %
Additional info is possible)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR