3-(Hydroxymethyl)indole 97%

Warning

Synonyms: Indole-3-methanol3-Indolcarbinol3-IndolmethanolIndole-3-carbinol(1H-Indol-3-yl)methanol(Indole-3-yl)methanol3-Indolemethanol

AC12219-0050 AC12219-0250
200025-570EA 67.8 USD
200025-570 200037-646
3-(Hydroxymethyl)indole 97%
3-(Hydroxymethyl)indole
Formula: C₉H₉NO
MW: 147.18 g/mol
Melting Pt: 92…94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005632
CAS Number: 700-06-1

Order Now

Specification Test Results

Appearance Off-white to yellow to light orange Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 92 to 100 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR