2-Imidazolidinone hemihydrate 99+%

Warning

Synonyms: 2-Imidazolidone hemihydrate

AC12204-0025 AC12204-0250 AC12204-5000
200041-092EA 232 USD
200041-092 200022-728 200041-094
2-Imidazolidinone hemihydrate 99+%
2-Imidazolidinone hemihydrate
Formula: C₃H₆N₂O·0.5H₂O
MW: 95.10 (86.09 anhy)
Boiling Pt: >100 °C (760 mmHg)
Melting Pt: 58 °C
Flash Pt: 265 °C
Storage Temperature: Ambient
MDL Number: MFCD02662355
CAS Number: 121325-67-5
EINECS: 204-436-4

Order Now

Specification Test Results

Appearance White to light yellow Adhering crystalline powder or crystals
Infrared spectrum Conforms
Water ≤13 %
GC ≥99.0 %
Elemental Analysis 5.17 to 5.55 % H
Elemental Analysis 5.1 to 5.6 % N

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR