2-Imidazolidinethione 98%

Danger

Synonyms: 2-ImidazolidinethioneEthlenethioureaEthylenethiourea2-Mercaptoimidazoline

AC12203-0010 AC12203-2500
200034-842EA 136 USD
200034-842 200022-726
2-Imidazolidinethione 98%
2-Imidazolidinethione
Formula: C₃H₆N₂S
MW: 102.16 g/mol
Melting Pt: 197…201 °C
Flash Pt: 252 °C
Storage Temperature: Ambient
MDL Number: MFCD00005276
CAS Number: 96-45-7
EINECS: 202-506-9
ADR: 3,II

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 196 to 200 °C
HPLC ≥97.5 %
Stabilizer Approx 500 ppm hydroquinone

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR