(±)-3-Hydroxytetrahydrofuran 97%

Warning

Synonyms: 3-HydroxytetrahydrofuranOxolan-3-ol

AC12194-0250 AC12194-0050
200000-550EA 109 USD
200000-550 200009-972
(±)-3-Hydroxytetrahydrofuran 97%
(±)-3-Hydroxytetrahydrofuran
Formula: C₄H₈O₂
MW: 88.1100 g/mol
Boiling Pt: 181 °C (760 mmHg)
Density: 1.09 g/cm³
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00005374
CAS Number: 453-20-3
EINECS: 207-219-2

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4510 (20 °C, 589 nm)
GC ≥96.0 %
UV at 260 nm A: ≤0.04
UV (0.25 M in water)
UV at 200 nm A: ≤0.3
UV at 210 nm A: ≤0.2
UV at 220 nm A: ≤0.1
UV at 230 nm A: ≤0.075
UV at 240 nm A: ≤0.05
Titration after Ion exchange ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR