2-Hydroxy-1,4-naphthoquinone 99%

Warning

Synonyms: 2-Hydroxy-p-naphthoquinone2-Hydroxy-1,4-naphoquinone

AC12163-1000
200028-788EA 125 USD
200028-788
2-Hydroxy-1,4-naphthoquinone 99%
2-Hydroxy-1,4-naphthoquinone
Formula: C₁₀H₆O₃
MW: 174.16 g/mol
Melting Pt: 192…195 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00001678
EINECS: 201-496-3

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR