4'-Hydroxy-2'-methylacetophenone 97%
|
Warning
|
Synonyms:
1-(4-Hydroxy-2-methylphenyl)ethanone
AC12139-0050
AC12139-0250
AC12139-1000
103509-608EA
472
USD
103509-608
103509-606
200010-452
4'-Hydroxy-2'-methylacetophenone 97%
4'-Hydroxy-2'-methylacetophenone
Formula:
C₉H₁₀O₂ MW: 150.18 g/mol Boiling Pt: 313 °C (760 mmHg) Melting Pt: 127…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002303 CAS Number: 875-59-2 EINECS: 212-874-2 |
Specification Test Results
Appearance | Off-white to beige to tan Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 127 to 132 °C |
HPLC | ≥96.0 % |
Color scale | ≤20 APHA |
Acidity/alkalinity | ≤0.00008 meq/g |
Substance darkened by H2SO4 | passes test |
Calcium (Ca) | ≤0.2 ppm |
Copper (Cu) | ≤0.02 ppm |
Iron (Fe) | ≤0.1 ppm |
Lead (Pb) | ≤0.02 ppm |
Magnesium (Mg) | ≤0.05 ppm |
Potassium (K) | ≤0.1 ppm |
Sodium (Na) | ≤0.1 ppm |
Zinc (Zn) | ≤0.1 ppm |
Hydrocarbons | ≤0.1 % Aromatic hydrocarbons |
Total phosphorus (P) | ≤0.1 ppm |
Total silicon (Si) | ≤0.05 ppm |
Total sulfur (S) | ≤10 ppm |
Bromine number | ≤0.5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...