4-Hydroxy-3-methoxybenzylamine hydrochloride 98%

Warning

Synonyms: 4-(Aminomethyl)-2-methoxyphenol hydrochloride

AC12137-0050 AC12137-0010
200047-924EA 27.4 USD
200047-924 103509-604
4-Hydroxy-3-methoxybenzylamine hydrochloride 98%
4-Hydroxy-3-methoxybenzylamine hydrochloride
Formula: C₈H₁₂ClNO₂
MW: 189.64 g/mol
Melting Pt: 219…221 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012864
CAS Number: 7149-10-2
EINECS: 230-468-3

Order Now

Specification Test Results

Appearance Off-white Powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR