N-(2-Hydroxyethyl)morpholine 99%

Warning

Synonyms: 2-Morpholinoethanol4-(2-Hydroxyethyl)morpholine

AC12118-5000 AC12118-0050 AC12118-1000
200013-456EA 56.5 USD
200013-456 200025-984 200019-880
N-(2-Hydroxyethyl)morpholine 99%
N-(2-Hydroxyethyl)morpholine
Formula: C₆H₁₃NO₂
Melting Pt: –1 °C
Density: 1.08 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00006180
CAS Number: 622-40-2
EINECS: 210-734-5

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4760 to 1.4780 (20 °C, 589 nm)
Water ≤0.5 %
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR