Humic acid sodium salt 45 - 70% (as humic acid), crystalline powder, Technical Grade

Warning

Synonyms: Sodium bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylate

AC12086-1000 AC12086-0050 AC12086-0010
200012-694EA 209 USD
200012-694 200025-542 200006-656
Humic acid sodium salt 45 - 70% (as humic acid), crystalline powder, Technical Grade
Humic acid sodium salt

Appearance: Black Crystalline powder

Formula: C₉H₈Na₂O₄
MW: 226.139 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00135560
CAS Number: 68131-04-4

Order Now

Specification Test Results

Appearance Black Crystalline powder
Infrared spectrum Conforms
Assay 45 to 70 % (as Humic acid)
Loss on drying 14 % to 25 % (105 °C, 4 h)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR