L-Histidine methyl ester dihydrochloride 97%

Warning

Synonyms: L(+)-Histidine methyl ester dihydrochlorideL-Methyl histidinate dihydrochlorideH-His-OMe.2HCl

AC12082-0050 AC12082-0250 AC12082-1000
200013-012EA 200 USD
200013-012 200019-500 103509-578
L-Histidine methyl ester dihydrochloride 97%
L-Histidine methyl ester dihydrochloride
Formula: C₇H₁₁N₃O₂
MW: 242.10 g/mol
Melting Pt: 207 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012701
CAS Number: 7389-87-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %
HPLC ≥96.0 %
Specific optical rotation +8° to +11° (20 °C, 589 nm) (c=2, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR