Gramine ≥99%

Danger

Synonyms: 3-(Dimethylaminomethyl)-Indole

AC12018-0100 AC12018-5000
103530-692EA 312 USD
103530-692 200031-486
Gramine ≥99%
Gramine
Formula: C₁₁H₁₄N₂
MW: 174.25 g/mol
Melting Pt: 131…139 °C
Flash Pt: 167 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005629
CAS Number: 87-52-5
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance Beige Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
Sulfated ash ≤0.5 %
HPLC ≥98.5 %
Titration with HClO4 ≥98.5 % (On anhydrous substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR