Glycine methyl ester hydrochloride (H-Gly-OMe.HCl) 98%

Synonyms: Methyl glycinate hydrochlorideH-Gly-OMe.HCl

AC12010-1000 AC12010-0025 AC12010-5000
103516-424EA 532.3 USD
103516-424 200006-586 200019-266
Glycine methyl ester hydrochloride (H-Gly-OMe.HCl) 98%
Glycine methyl ester hydrochloride (H-Gly-OMe.HCl)
Formula: C₃H₈ClNO₂
MW: 125.56 g/mol
Melting Pt: 173…177 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012870
CAS Number: 5680-79-5
EINECS: 227-139-1

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration with HClO4 97.5 to 102.5 %
GC ≥95 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR