DL-Glutamic acid monohydrate 98+%
Synonyms:
DL-Glutamic Acid Monohydrate
AC11993-0010
AC11993-1000
200018-934EA
359
USD
200018-934
200018-936
DL-Glutamic acid monohydrate 98+%
DL-Glutamic acid monohydrate
Formula:
C₅H₉NO₄·1H₂O MW: 165.15 g/mol (147.13 anhy) Melting Pt: 194 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00150703 EINECS: 210-522-2 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Water | ≤15.5 % (K.F.) |
Titration with NaOH | ≥98.0 % |
Specific optical rotation | +31° to +35° (20 °C, 589 nm) (c=1, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...