Furfurylamine 99+%
|
Danger
|
Synonyms:
2-(Aminomethyl)furan
AC11980-0010
AC11980-2500
AC11980-0050
200031-440EA
172
USD
200031-440
200031-438
200019-224
Furfurylamine 99+%
Furfurylamine
Formula:
C₅H₇NO MW: 97.12 g/mol Boiling Pt: 145…146 °C (761 mmHg) Melting Pt: –70 °C Density: 1.09 g/cm³ Flash Pt: 37 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003258 CAS Number: 617-89-0 EINECS: 210-536-9 UN: 2526 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to yellow-brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4890 to 1.4910 (20 °C, 589 nm) |
GC | ≥99.0 % |
Titration with HClO4 | ≥99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...