Piperidine-N-carbaldehyde 99%
|
Danger
|
Synonyms:
N-Formylpiperidine
, 1-Formylpiperidine
AC11973-2500
AC11973-0010
200020-052EA
72.5
USD
200020-052
200038-196
Piperidine-N-carbaldehyde 99%
Piperidine-N-carbaldehyde
Formula:
C₆H₁₁NO MW: 113.16 g/mol Melting Pt: –31 °C Density: 1.019 g/cm³ Flash Pt: 91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006483 CAS Number: 2591-86-8 EINECS: 219-986-0 UN: 2810 ADR: 6.1,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4830 to 1.4850 (20 °C, 589 nm) |
GC | ≥98.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...