Formamidine acetate 99%
|
Warning
|
AC11971-1000
AC11971-5000
200031-418EA
186
USD
200031-418
200012-536
Formamidine acetate 99%
Formamidine acetate
Formula:
C₃H₈N₂O₂ Melting Pt: 157…161 °C Density: 1.124 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00012866 CAS Number: 3473-63-0 EINECS: 222-442-5 |
Specification Test Results
Appearance | White to light yellow Crystalline powder or crystals |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥98.5 % |
GC | ≥97.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...