FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) 90%, pure
|
Danger
|
Synonyms:
5-isothiocyanatofluorescein
, 5-FITC
, Fluorescein Isothiocyanate, Isomer I
, Fluorescein Isothiocyanate
, 3',6'-Dihydroxy-5-isothiocyanato-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one
AC11925-2500
200000-484EA
66.3
USD
200000-484
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) 90%, pure
FITC (Fluorescein 5-isothiocyanate, Fluorescein isothiocyanate isomer I)
Formula:
C₂₁H₁₁NO₅S MW: 389.381 g/mol Melting Pt: >360 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005063 CAS Number: 3326-32-7 EINECS: 222-042-0 |
Specification Test Results
Appearance | Orange Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥88 % |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...